Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM92
Entrez Gene ID
507710
Gene Name
transmembrane protein 92
Gene Aliases
-
Chromosome Location
Chr.19: 36388022 - 36396056 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083651.2 NP_001077120.1 2 - 3 148 98
GenBank mRNA BC134690.1 - 2 - 3 149 98

View Full Product Details