Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM92
Entrez Gene ID
507710
Gene Name
transmembrane protein 92
Gene Aliases
-
Chromosome Location
Chr.19: 36388022 - 36396056 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083651.2 NP_001077120.1 3 - 4 244 100
GenBank mRNA BC134690.1 - 3 - 4 245 100

View Full Product Details