Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSL
Entrez Gene ID
515200
Gene Name
cathepsin L
Gene Aliases
CTSL1
Chromosome Location
Chr.8: 80974569 - 80980418 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083686.2 NP_001077155.1 5 - 6 704 76
GenBank mRNA BC134741.1 - 5 - 6 706 76

View Full Product Details