Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNB2
Entrez Gene ID
281202
Gene Name
G protein subunit beta 2
Gene Aliases
-
Chromosome Location
Chr.25: 35909868 - 35915046 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097561.1 NP_001091030.1 1 - 2 52 139
GenBank mRNA BC140489.1 - 1 - 2 52 139

View Full Product Details