Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNB2
Entrez Gene ID
281202
Gene Name
G protein subunit beta 2
Gene Aliases
-
Chromosome Location
Chr.25: 35909868 - 35915046 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097561.1 NP_001091030.1 8 - 9 841 90
GenBank mRNA BC140489.1 - 8 - 9 841 90
M16480.1 - 7 - 8 660 90
M16539.1 - 3 - 4 397 90
M36431.1 - 7 - 8 660 90

View Full Product Details