Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASLG
Entrez Gene ID
407111
Gene Name
Fas ligand
Gene Aliases
-
Chromosome Location
Chr.16: 40140582 - 40171582 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098859.2 NP_001092329.1 1 - 2 524 72
GenBank mRNA BC146056.1 - 1 - 2 526 72
JN380921.1 - 1 - 2 335 72

View Full Product Details