Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP3
Entrez Gene ID
281610
Gene Name
A-kinase anchoring protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 105495961 - 105523869 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101039.2 NP_001094509.1 4 - 5 2618 74
GenBank mRNA AF093407.1 - 2 - 3 2401 74
BC150040.1 - 4 - 5 2620 74

View Full Product Details