Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLBD1
Entrez Gene ID
317710
Gene Name
phospholipase B domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 95273631 - 95362983 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101044.2 NP_001094514.2 7 - 8 1252 104
NM_001166298.1 NP_001159770.1 9 - 10 1594 104
GenBank mRNA AJ299408.1 - 9 - 10 1299 104
BC149435.1 - 7 - 8 1238 104

View Full Product Details