Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1A
Entrez Gene ID
282031
Gene Name
RAP1A, member of RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 31588106 - 31676111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174448.5 NP_776873.1 6 - 7 616 139
GenBank mRNA BC134566.1 - 6 - 7 546 139
DR697686.1 - 2 - 3 158 139
J04196.1 - 5 - 6 469 139

View Full Product Details