Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC22
Entrez Gene ID
522691
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.3: 91489778 - 91514089 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098055.1 NP_001091524.1 2 - 3 710 71
GenBank mRNA BC140508.1 - 2 - 3 710 71

View Full Product Details