Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX26
Entrez Gene ID
508945
Gene Name
testis expressed 26
Gene Aliases
C12H13orf26
Chromosome Location
Chr.12: 29967342 - 29991507 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098913.2 NP_001092383.1 1 - 2 87 69
GenBank mRNA BC146109.1 - 1 - 2 88 69

View Full Product Details