Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBLN2
Entrez Gene ID
509847
Gene Name
cerebellin 2 precursor
Gene Aliases
-
Chromosome Location
Chr.24: 5330841 - 5339281 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098919.1 NP_001092389.1 3 - 4 669 64
GenBank mRNA BC142191.1 - 3 - 4 669 64

View Full Product Details