Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC43
Entrez Gene ID
513649
Gene Name
coiled-coil domain containing 43
Gene Aliases
-
Chromosome Location
Chr.19: 44427059 - 44436411 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098962.2 NP_001092432.1 2 - 3 341 81
GenBank mRNA BC142025.1 - 2 - 3 342 81

View Full Product Details