Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC31A1
Entrez Gene ID
518976
Gene Name
solute carrier family 31 member 1
Gene Aliases
-
Chromosome Location
Chr.8: 102470399 - 102504615 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100381.1 NP_001093851.1 3 - 4 311 142
GenBank mRNA BC150134.1 - 3 - 4 311 142

View Full Product Details