Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RESF1
Entrez Gene ID
510651
Gene Name
retroelement silencing factor 1
Gene Aliases
C5H12orf35, KIAA1551
Chromosome Location
Chr.5: 77923726 - 77950979 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101072.2 NP_001094542.1 5 - 6 5359 96
GenBank mRNA BC151612.1 - 5 - 6 5360 96

View Full Product Details