Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIFC1
Entrez Gene ID
510995
Gene Name
kinesin family member C1
Gene Aliases
-
Chromosome Location
Chr.23: 7569904 - 7585473 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101936.2 NP_001095406.1 4 - 5 431 72
GenBank mRNA BC151443.1 - 4 - 5 432 72

View Full Product Details