Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A18
Entrez Gene ID
514287
Gene Name
membrane spanning 4-domains A18
Gene Aliases
-
Chromosome Location
Chr.29: 37085870 - 37103721 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101992.2 NP_001095462.1 4 - 5 708 62
GenBank mRNA BC149291.1 - 4 - 5 709 62

View Full Product Details