Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC18C
Entrez Gene ID
516050
Gene Name
C-type lectin domain family 18 member C
Gene Aliases
MRCL
Chromosome Location
Chr.18: 1831608 - 1845295 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102034.1 NP_001095504.1 4 - 5 629 59
GenBank mRNA BC149637.1 - 4 - 5 629 59

View Full Product Details