Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPC5
Entrez Gene ID
522828
Gene Name
glypican 5
Gene Aliases
-
Chromosome Location
Chr.12: 65755006 - 67339141 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102070.1 NP_001095540.1 4 - 5 1493 118
GenBank mRNA BC151393.1 - 4 - 5 1493 118

View Full Product Details