Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
520842
Gene Name
Kruppel like factor 4
Gene Aliases
-
Chromosome Location
Chr.8: 97174495 - 97179140 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105385.1 NP_001098855.1 2 - 3 1087 79
GenBank mRNA BC134523.1 - 2 - 3 1087 79

View Full Product Details