Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLA2G4B
Entrez Gene ID
505485
Gene Name
phospholipase A2, group IVB (cytosolic)
Gene Aliases
-
Chromosome Location
Chr.10: 37228285 - 37239358 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114657.1 NP_001108129.1 9 - 10 787 155

View Full Product Details