Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHCBP1L
Entrez Gene ID
528041
Gene Name
SHC binding and spindle associated 1 like
Gene Aliases
C16H1ORF14
Chromosome Location
Chr.16: 63941657 - 63985594 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046290.2 NP_001039755.2 9 - 10 1745 124
GenBank mRNA BC112556.1 - 9 - 10 1746 124
DT857793.1 - 3 - 4 480 124

View Full Product Details