Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM71F2
Entrez Gene ID
540061
Gene Name
family with sequence similarity 71 member F2
Gene Aliases
-
Chromosome Location
Chr.4: 92648962 - 92674006 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046467.3 NP_001039932.2 1 - 2 244 70

View Full Product Details