Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDB2
Entrez Gene ID
617976
Gene Name
LIM domain binding 2
Gene Aliases
-
Chromosome Location
Chr.6: 111698867 - 112153623 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046611.3 NP_001040076.2 4 - 5 746 95
NM_001314058.1 NP_001300987.1 4 - 5 746 95
GenBank mRNA BC105321.1 - 4 - 5 616 95
DV882781.1 - 4 - 5 419 95
EV635520.1 - 4 - 5 734 95

View Full Product Details