Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETD6
Entrez Gene ID
539651
Gene Name
SET domain containing 6, protein lysine methyltransferase
Gene Aliases
-
Chromosome Location
Chr.18: 26299633 - 26304394 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098102.1 NP_001091571.1 2 - 3 177 81
GenBank mRNA BC140545.1 - 2 - 3 177 81

View Full Product Details