Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLPH3L
Entrez Gene ID
532555
Gene Name
golgi phosphoprotein 3 like
Gene Aliases
-
Chromosome Location
Chr.3: 20037044 - 20085018 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099052.1 NP_001092522.1 2 - 3 321 66
GenBank mRNA BC146227.1 - 2 - 3 321 66

View Full Product Details