Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDE2
Entrez Gene ID
534664
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C16H1orf55
Chromosome Location
Chr.16: 29005211 - 29020714 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099065.2 NP_001092535.1 6 - 7 1153 128
GenBank mRNA BC142453.1 - 6 - 7 1155 128

View Full Product Details