Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD1B
Entrez Gene ID
529700
Gene Name
regulation of nuclear pre-mRNA domain containing 1B
Gene Aliases
C13H20ORF77
Chromosome Location
Chr.13: 66909647 - 66967484 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101134.1 NP_001094604.1 4 - 5 910 79
GenBank mRNA BC148107.1 - 4 - 5 910 79

View Full Product Details