Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOX5
Entrez Gene ID
531303
Gene Name
NADPH oxidase 5
Gene Aliases
-
Chromosome Location
Chr.10: 15827976 - 15890483 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101137.1 NP_001094607.1 10 - 11 1659 90

View Full Product Details