Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM266
Entrez Gene ID
536930
Gene Name
transmembrane protein 266
Gene Aliases
C21H15orf27
Chromosome Location
Chr.21: 31337017 - 31461073 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101168.1 NP_001094638.1 9 - 10 940 89
GenBank mRNA BC133555.1 - 6 - 7 633 89
BC148140.1 - 9 - 10 940 89

View Full Product Details