Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJA4
Entrez Gene ID
528615
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
-
Chromosome Location
Chr.21: 30653151 - 30669872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102120.2 NP_001095590.2 3 - 4 574 98
GenBank mRNA BC110008.1 - 3 - 4 350 98
BC151654.1 - 3 - 4 366 98
DT834391.1 - 3 - 4 574 98

View Full Product Details