Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAPBPL
Entrez Gene ID
529141
Gene Name
TAP binding protein like
Gene Aliases
TAPBP-R
Chromosome Location
Chr.5: 103922266 - 103929878 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102121.2 NP_001095591.1 5 - 6 1607 80
GenBank mRNA BC148877.1 - 5 - 6 1608 80

View Full Product Details