Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP38
Entrez Gene ID
534262
Gene Name
ubiquitin specific peptidase 38
Gene Aliases
-
Chromosome Location
Chr.17: 14684121 - 14716484 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102167.1 NP_001095637.1 2 - 3 1305 80
GenBank mRNA BC148040.1 - 2 - 3 1305 80

View Full Product Details