Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NSUN5
Entrez Gene ID
616714
Gene Name
NOP2/Sun RNA methyltransferase 5
Gene Aliases
NSUN5C
Chromosome Location
Chr.25: 33820199 - 33825835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102303.2 NP_001095773.1 1 - 2 114 68
GenBank mRNA BC149960.1 - 1 - 2 117 68

View Full Product Details