Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P3H1
Entrez Gene ID
539976
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
LEPRE1
Chromosome Location
Chr.3: 103472286 - 103491713 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103291.1 NP_001096761.1 1 - 2 489 67
GenBank mRNA BC123438.1 - 1 - 2 489 67

View Full Product Details