Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSBP3
Entrez Gene ID
530184
Gene Name
single stranded DNA binding protein 3
Gene Aliases
-
Chromosome Location
Chr.3: 91841572 - 92007897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104986.1 NP_001098456.1 3 - 4 321 115
GenBank mRNA BC151395.1 - 3 - 4 321 115

View Full Product Details