Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLN6
Entrez Gene ID
617615
Gene Name
CLN6 transmembrane ER protein
Gene Aliases
-
Chromosome Location
Chr.10: 15050676 - 15067684 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109984.1 NP_001103454.1 5 - 6 545 85

View Full Product Details