Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAQ
Entrez Gene ID
536654
Gene Name
G protein subunit alpha q
Gene Aliases
-
Chromosome Location
Chr.8: 53685191 - 53995695 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110002.1 NP_001103472.1 1 - 2 361 87

View Full Product Details