The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113230.1 | NP_001106701.1 | 3 - 4 | 258 | 66 |
GenBank mRNA | BC112758.1 | - | 3 - 4 | 258 | 66 |