Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD6
Entrez Gene ID
616226
Gene Name
COMM domain containing 6
Gene Aliases
-
Chromosome Location
Chr.12: 50570857 - 50599403 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113312.2 NP_001106783.1 3 - 4 235 149
GenBank mRNA BC112466.1 - 3 - 4 236 149

View Full Product Details