Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPX
Entrez Gene ID
618018
Gene Name
centromere protein X
Gene Aliases
BHLHB2, STRA13
Chromosome Location
Chr.19: 50887829 - 50891455 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114527.2 NP_001107999.1 1 - 2 58 56
GenBank mRNA BC111642.1 - 1 - 2 59 56

View Full Product Details