Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEK2
Entrez Gene ID
784941
Gene Name
pleckstrin 2
Gene Aliases
-
Chromosome Location
Chr.10: 79278720 - 79299439 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102334.1 NP_001095804.1 3 - 4 399 130
GenBank mRNA BC148042.1 - 3 - 4 399 130

View Full Product Details