Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPIND1
Entrez Gene ID
100125764
Gene Name
serpin family D member 1
Gene Aliases
-
Chromosome Location
Chr.17: 72232049 - 72238750 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105046.2 NP_001098516.1 2 - 3 971 71
GenBank mRNA BC149414.1 - 2 - 3 972 71

View Full Product Details