Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF75A
Entrez Gene ID
100126179
Gene Name
zinc finger protein 75a
Gene Aliases
-
Chromosome Location
Chr.25: 2753806 - 2765657 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109965.1 NP_001103435.1 6 - 7 1255 122
GenBank mRNA BC149191.1 - 6 - 7 1255 122

View Full Product Details