Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2D14
Entrez Gene ID
282211
Gene Name
cytochrome P450, family 2, subfamily D, polypeptide 6
Gene Aliases
CYP2D6
Chromosome Location
Chr.5: 113087282 - 113094933 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174529.2 NP_776954.1 4 - 5 716 151
GenBank mRNA BC149952.1 - 4 - 5 902 151
X68481.1 - - 716 151

View Full Product Details