Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GNLY
- Entrez Gene ID
-
404173
- Gene Name
- granulysin
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- LOC104968634
- Entrez Gene ID
-
104968634
- Gene Name
- antimicrobial peptide NK-lysin
- Gene Aliases
-
NK2B, NK2C
- Chromosome Location
-
Chr.11: 49225782 - 49228290 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC114176.1 |
- |
5 - 5 |
548 |
153 |
- Gene Symbol
- LOC112448791
- Entrez Gene ID
-
112448791
- Gene Name
- antimicrobial peptide NK-lysin-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 49258428 - 49260931 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC114178.1 |
- |
5 - 5 |
547 |
153 |
View Full Product Details