Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC184
Entrez Gene ID
617289
Gene Name
coiled-coil domain containing 184
Gene Aliases
C5H12orf68
Chromosome Location
Chr.5: 32109728 - 32111603 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078073.1 NP_001071541.1 1 - 1 1750 147
GenBank mRNA BC120392.1 - 1 - 1 1750 147

View Full Product Details