Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL5
Entrez Gene ID
514285
Gene Name
angiopoietin like 5
Gene Aliases
-
Chromosome Location
Chr.15: 6889264 - 6906335 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128069.1 NP_001121541.1 5 - 6 792 63
GenBank mRNA EU599228.1 - 5 - 6 792 63

View Full Product Details