Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TSC22D3
Entrez Gene ID
100125309
Gene Name
TSC22 domain family member 3
Gene Aliases
-
Chromosome Location
Chr.X: 56329795 - 56396388 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103342.1 NP_001096812.1 3 - 4 543 73
GenBank mRNA BC148873.1 - 3 - 4 543 73
BC148966.1 - 2 - 3 226 73

View Full Product Details