Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGFBR2
Entrez Gene ID
535376
Gene Name
transforming growth factor beta receptor 2
Gene Aliases
-
Chromosome Location
Chr.22: 5091294 - 5183754 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159566.2 NP_001153038.1 4 - 5 1486 58

View Full Product Details